missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisobutylaluminium chloride, 0.8M solution in heptane, AcroSeal™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid

Specifications
Specifications
Density | 0.7260g/mL |
Flash Point | −4°C |
Packaging | AcroSeal™ Glass bottle |
Quantity | 800 mL |
Specific Gravity | 0.726 |
Solubility Information | Solubility in water: vigorous reaction. Other solubilities: miscible with saturated aliphatic and aromatic,hydrocarbons |
Formula Weight | 176.67 |
Chemical Name or Material | Diisobutylaluminium chloride |
Safety and Handling





Product Identifier
- Diisobutylaluminium chloride, 0.6M solution in heptane
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 1
- H304-May be fatal if swallowed and enters airways.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
- H336-May cause drowsiness or dizziness.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P331-Do NOT induce vomiting.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Diisobutylaluminium chloride, 0.8M solution in heptane, AcroSeal™ >
Spot an opportunity for improvement?Share a Content Correction