missing translation for 'onlineSavingsMsg'
Learn More

Scharlab DIMETYLSULFOXID PA 1L

Brand:  Scharlab SU01531000

DIMETYLSULFOXID PA 1L

Product Code. 11354044

  • 1075.00 DKK / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Scharlab DIMETYLSULFOXID PA 1L >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.