Learn More
Thermo Scientific™ Dionex™ MAbPac™SCX-10 Ion Exchange Columns
For the high-resolution separation and analysis of monoclonal antibody variants
Brand: Thermo Scientific™ 088782
This item is not returnable.
View return policy
Description
- Unequalled high-resolution separations
- Exceptionally high efficiency
- Highest resolution of monoclonal antibody variants
- Unmatched column-to-column and lot-to-lot reproducibility
- Hydrophobic interactions eliminated
- Excellent for stability testing
Specifications
Bio-LC | |
1 Lot of Resin |
Product Content Correction
Product Title
Thermo Scientific™ Dionex™ MAbPac™SCX-10 Ion Exchange Columns > Length: 250mm; ID: 4mm; 1 Lot of Resin
Spot an opportunity for improvement?Share a Content Correction