Learn More
Thermo Scientific™ Dionex™ PEEK Sleeves
Thermo Scientific™ Dionex PEEK Sleeves are precision cut and polished.
Brand: Thermo Scientific™ 161405
674.17 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Thermo Scientific™ Dionex PEEK Sleeves are precision cut and polished.
Specifications
For microtight union; 360 um OD | |
nanoLC | |
360 μm |
Connections with Microtight Union | |
Tubing Sleeve |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 9
Product Content Correction
Product Title
Thermo Scientific™ Dionex™ PEEK Sleeves > For connections with microtight union; 360um O.D.
Spot an opportunity for improvement?Share a Content Correction