Learn More
Diphenyliodonium hexafluorophosphate, 97%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 140.0°C to 144.0°C |
Assay Percent Range | 96% min. (HPLC) |
Packaging | Plastic bottle |
Linear Formula | (C6H5)2I+PF6- |
Quantity | 25 g |
Solubility Information | Solubility in water: .. Other solubilities: soluble in methanol |
Percent Purity | 97% |
Chemical Name or Material | Diphenyliodonium hexafluorophosphate |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0438855 | Certificate of Analysis | 25/08/2023 | 450910250,450911000,450910050 |
A0394448 | Certificate of Analysis | 14/12/2022 | 450910250,450911000,450910050 |
A0395852 | Certificate of Analysis | 14/12/2022 | 450910250,450911000,450910050 |
A0413722 | Certificate of Analysis | 13/12/2022 | 450910250,450911000,450910050 |
A0438509 | Certificate of Analysis | 27/01/2022 | 450910250,450911000,450910050 |
Safety and Handling

- Diphenyliodonium hexafluorophosphate
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.