missing translation for 'onlineSavingsMsg'
Learn More
Diquat Dibromide Monohydrate, SPEX CertiPrep™ Product Code.: 15206156

Diquat Dibromide Monohydrate, SPEX CertiPrep™

Product Code. 15206156 Shop All SPEX Certiprep Products
1 mL
Click to view available options
Quantity:
1 mL
This item is not returnable. View return policy

Product Code. 15206156

Brand: SPEX Certiprep S1752

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Diquat dibromide monohydrate6385-62-21000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

CAS 6385-62-2, 67-56-1
Packaging 2mL amber prescored glass ampule
CAS Min % 0.1, 99.9
Quantity 1 mL
CAS Max % 0.1, 99.9
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
Molecular Formula C12H14Br2N2O
MDL Number MFCD00211319
InChI Key KLJOSQAQZMLLMB-UHFFFAOYSA-L
SMILES O.[Br-].[Br-].C1C[N+]2=CC=CC=C2C2=CC=CC=[N+]12
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
IUPAC Name 7λ5,10λ5-diazatricyclo[8.4.0.02,7]tetradeca-1(14),2,4,6,10,12-hexaene-7,10-bis(ylium) hydrate dibromide
Molecular Weight (g/mol) 362.07
PubChem CID 115268
Type Certified Reference Material
Solvent or Matrix Methanol-P&T
Chemical Name or Material Diquat dibromide monohydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : methanol, diquat dibromide monohydrate

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diquat Dibromide Monohydrate, SPEX CertiPrep™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.