missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Direct Thermal Printable Labels, 25 mm Core

Printable paper labels for thermal transfer printers.

Brand:  Brady™ 236749

 View more versions of this product

Product Code. 11573135

  • 5360.00 DKK / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Direct thermal paper
  • Ideal for warehouse marking
Specifications

Specifications

Not Autoclavable
White
Direct Thermal Paper
Non-sterile
BBP™11 Label Printer, BBP™12 Industrial Label Printer, THT 76 mm Core Printers
3000 Labels
0.099 mm
101.6 mm
Permanent Acrylic
Matte
Direct Thermal
Warehouse Marking
1
Rectangle
155.6 mm
152.4 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers