Learn More
Uvex™ Dispenser blue/clear for Uvex disposable ear plugs Uvex
Brand: Uvex™ 2112000
244.38 DKK valid until 2025-06-08
Use promo code "24131" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Regular | |
No | |
Dispenser | |
Yes | |
Plastic | |
Wall Mountable |
na | |
Oval | |
Reusable | |
No | |
one2click Dispenser | |
No |
Promotions
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
Product Content Correction
Product Title
Uvex™ Dispenser blue/clear for Uvex disposable ear plugs Uvex >
Spot an opportunity for improvement?Share a Content Correction