Learn More
Miele Professional™ Dosing system
Dispenser module DOS K 85/1 for 5 litre canister with level indicator for the automatic dispensing of liquid cleaning agents
Brand: Miele Professional™ 9961080
Additional Details : Weight : 2.85000kg
Description
- Dispenser monitoring – in accordance with DIN EN ISO 15883
- For dispensing liquid cleaning agent or neutralising agent
- For dispensing chemical disinfectant
- 200 mm siphon for 5 litre containers
Specifications
Dispensing System | |
Washer-disinfectors, laboratory |