Learn More
Fisherbrand™ Disposable Powder-free Non-sterile Extra Protection Nitrile Examination Gloves
Resists a wide range of chemicals and resistant to puncture. Fisherbrand™ Disposable Powder-free Non-sterile Extra Protection Nitrile Examination Gloves are latex free.
92.30 DKK - 109.00 DKK
Specifications
Certifications/Compliance | Conforms to EN 455 parts 1 and 2 |
---|---|
Color | Purple |
Disposable | Single-use |
Latex Free | Yes |
Material | Nitrile |
Description
- 100% nitrile-resistant to a wide range of chemicals and three times more puncture resistant than latex
- Ergonomically designed with textured fingertips for improved grip
- Latex, protein and powder free to eliminate Type I allergies
- 0.10mm thickness
- Non-detectable residual accelerators
- Exceeds the requirements of EN 374 PPE Category III
- Conforms to EN 455 parts 1 and 2
Specifications
Conforms to EN 455 parts 1 and 2 | |
Single-use | |
Nitrile | |
Nitrile Examination Glove | |
0.10 mm | |
50/Pk. | |
30 cm |
Purple | |
Yes | |
Powder Free | |
Non-sterile | |
Textured | |
Puncture, wide range of chemicals |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Disposable Powder-free Non-sterile Extra Protection Nitrile Examination Gloves