Learn More
Greiner Bio-One Disposal Bags
For hygienic disposal of contaminated laboratory items. Greiner Bio-One Disposal Bags are available in polypropylene and polyamide.
Brand: Greiner Bio-One 644401
This item is not returnable.
View return policy
Description
- Non-sterile
- Polypropylene (PP) suitable for steam autoclaves
- Polyamide (PA) suitable for hot-air sterilisers
- Foil thickness: 0.05mm
- Nominal capacities from 10L to 130L
- For single use only
Protect from direct light
Specifications
Autoclavable | |
Translucent | |
Polyamide | |
780 x 400mm |
30 L | |
Hygienic disposal of contaminated laboratory items | |
0.05 mm |
Product Content Correction
Product Title
Greiner Bio-One Disposal Bags > Polyamide; Suitable for hot-air sterilisers; L x W: 780 x 400mm; Capacity: 30L
Spot an opportunity for improvement?Share a Content Correction