Learn More
Stainless Steel Dissecting Set
Use this kit for general laboratory applications. S Murray™ Stainless Steel Dissecting Set are suitable for theatre, ward, veterinary, dental and chiropody use.
Brand:  S Murray SAMCO Y261/73
Product Code. 12321379
  • 533.00 DKK / Each
Estimated Shipment: 05-06-2025
to see stock.
Explore more special offers
Description
  • Manufactured from high grade stainless steel
  • Complies with ISO 7153-1 specifications
  • Standard instruments are finished to a bright mirror polish
Specifications
Specifications
Stainless Steel
15 Ea.
Dissecting Set Wallet
Product Title
Stainless Steel Dissecting Set > Material: Stainless Steel