Learn More
Bochem™ Dissection Needle
Bochem™ Dissection Needle has a lancet tip and Stainless Steel handle.
Brand:  Bochem™ 12020
Product Code. 10020891
  • 63.80 DKK / Each
Estimated Shipment: 03-06-2025
to see stock.
Explore more special offers
Description
  • Stainless Steel
  • Lancet tip
Specifications
Specifications
Stainless Steel
Lancet
140 mm
Documents
One moment while we fetch your results.
Product Title
Bochem™ Dissection Needle > Length: 140mm