missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasintrumente Distilling Links with Socket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 05047002
This item is not returnable.
View return policy
Description
- Distilling links without condenser
- With two vertical NS cones
- With socket
- NS 14/23 for distillation thermometers of 55 mm built-in length
- Made of DURAN™ tubing
Specifications
200 mm | |
Duran Glass | |
Cone NS 19/26, Socket NS 14/23 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lenz Laborglasintrumente Distilling Links with Socket > 200 mm
Spot an opportunity for improvement?Share a Content Correction