missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey anti-Mouse IgG (H+L) Secondary Antibody, PE/Cy7, Novus Biologicals™
Donkey Polyclonal Antibody
Brand: Novus Biologicals NB120-6707PECY7
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Fluorophore-linked immunosorbent assay
Specifications
IgG (H+L) | |
Polyclonal | |
PBS | |
Mouse IgG whole molecule | |
500 ÎĽg | |
Adaptive Immunity, Immunology | |
Mouse | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Fluorophore-Linked Immunosorbent Assay | |
PE-Cyanine7 | |
Donkey | |
Immunogen affinity purified | |
RUO | |
Secondary | |
Store at 4°C in the dark. Do not freeze. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Donkey anti-Mouse IgG (H+L) Secondary Antibody, PE/Cy7, Novus Biologicals™ > 500 μg; PE-Cyanine7
Spot an opportunity for improvement?Share a Content Correction