Learn More
Kartell™ Double Ball with Net
Suitable for automatic burettes and flashchromatography columns
Brand:  Kartell™ 0391200
362.90 DKK valid until 2025-07-13
Use promo code "24135" to get your promotional price.
 View more versions of this product
Product Code. 10594621
  • 382.00 DKK / Each
Elevate Your Beverage Testing with Precision Solutions
Use promo code 24135 at checkout
View Available Promotions
Only null left
Estimated Shipment: 23-07-2025
Specifications
Red rubber
46 mm
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Kartell™ Double Ball with Net > 46mm O.D.