Learn More
Bochem™ Double Ended Micro Spatula
Bochem™ Double Ended Micro Spatula for handling and sampling small samples.
Brand:  Bochem™ 3029
 View more versions of this product
Product Code. 10030881
  • 18.75 DKK / Each
Estimated Shipment: 27-02-2025
to see stock.
Explore more special offers
Description
  • Double ended
  • Stainless Steel
Specifications
Specifications
Double Ended
50 mm
Stainless Steel (18/10)
Micro Spatula
9 mm
185 mm
Product Title
Bochem™ Double Ended Micro Spatula > Length: 185mm