Learn More
Dräger™ CMS Chips: Sulfur Dioxide
Suitable for 10 measurements of one gas
1660.00 DKK
Product Code | Brand | Type | Measurement Range | Response Time | Humidity Range | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Measurement Range | Response Time | Humidity Range | Price | Quantity & Availability | ||
12304649
|
Dräger™
6406180 |
Sulfur Dioxide CMS Chip | 5 to 150 | 40 to 360 sec. | 1 to 40mg/L |
1660.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.Specifications
700 to 1100 hpa | |
Sulfur Dioxide CMS Chip |