Learn More
Dräger™ Diffusion Tubes with Direct Indication
Convenient, long-term TWA monitoring—direct reading, with no lab analysis required
1225.00 DKK - 1630.00 DKK
Specifications
Quantity 10
Response Time 8 hr.
View More Specs
Products 3
Product Code Brand Humidity Range Measurement Range Temperature Range (English) Temperature Range (Metric) Price Quantity & Availability  
Product Code Brand Humidity Range Measurement Range Temperature Range (English) Temperature Range (Metric) Price Quantity & Availability  
10464482
View Documents
Dräger™
6733111
3mg/L 1.3 to 25 64.4°F to 71.6°F 18° to 22°C This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12782507
View Documents
Dräger™
8101301
1 to 16mg/L 2.5 to 188 32°F to 104°F 0°C to 40°C
1630.00 DKK
Pack of 10
10474482
View Documents
Dräger™
6733091
Less than15mg/L 1.3 to 38 32°F to 104°F 0°C to 40°C
1610.00 DKK
Each
Description
Description
Tubes sample ambient air by gaseous diffusion with no pumps or moving parts. To use, break off the tube end with the special holder, put the tube in the holder, and clip it to your pocket or lapel.
  • Obtain time-weighted average (TWA) measurements of exposure to toxic gases for periods of up to 8 hours
  • Reagent changes color as it reacts with the airborne contaminant
  • Final results are calculated by dividing the tube reading by the number of hours it was used
  • Designed and manufactured using ISO 90001:2000-certified processes
Order Info
Use with accessory clip-on holder (Cat. No. 17-986-4; available separately below). Ten tubes per pack.
Specifications
Specifications
10
8 hr.