Learn More
Draeger Short Term Tubes
Easy to use Draeger tubes are proven to be very cost effective and reliable method for th measurement of gases.
Brand: Draeger 8103741
This item is not returnable.
View return policy
Description
- Quick and targeted measurements
- Easy handling either manually using the Draeger-Tube pump accuro or automatically with the Draeger X-act 5000
- Field of application includes air examinations at the workplace, technical gas analysis in areas of emission/immission concentrations, measurement of compressed air/gases Measures a large number of different gases and vapours
Specifications
i-Propanol 50/a | |
Short-term Draeger Sampling Tubes | |
Spot Measurement of More That 500 Different Gases |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Draeger Short Term Tubes > Gases Measured: i-Propanol 50/a, Short-term Draeger sampling tubes
Spot an opportunity for improvement?Share a Content Correction