Learn More
Kern Sohn Draft Shield Rear Panel
Draft shield rear panel with integrated ioniser to neutralise electrostatic charge.
Brand:  Kern Sohn ABP-A01
Encompass_Preferred
 View more versions of this product
Product Code. 17351513
  • 15133.73 DKK / Each
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Fitted in place of the existing glass rear panel of the draft shield
  • Includes rear panel, ionizer, and power supply
Specifications
Specifications
ABP Range Balances
Plastic
Rear Panel, Ionizer, and Power Supply