Learn More
Kern Sohn Draft Shield Rear Panel
Draft shield rear panel with integrated ioniser to neutralise electrostatic charge.
Brand: Kern Sohn ALJ-A03
This item is not returnable.
View return policy
Description
- Fitted in place of the existing glass rear panel of the draft shield
- Includes rear panel, ionizer, and power supply
Specifications
ALJ Range Balances | |
Plastic | |
Rear Panel, Ionizer, and Power Supply |