Learn More
Lenz Laborglasinstrumente™ Dropping funnel, conical-shaped, with PTFE plug
Made of borosilicate glass. Lenz Laborglasinstrumente™ Dropping funnel, conical-shaped, with PTFE plug are used for transferring fluids. The funnel is fitted with a stopcock which allows the flow to be controlled.
Brand:  Lenz Laborglasinstrumente™ 04477004
 View more versions of this product
Product Code. 11826214
  • 1155.00 DKK / Each
Estimated Shipment: 27-05-2025
to see stock.
Explore more special offers
Description
  • Conical shape with NS sleeve and NS core
  • With PTFE plug
  • With threaded valve protection
  • Borosilicate glass
Specifications
Specifications
1000 mL
Borosilicate Glass
6.0 mm bore
PTFE
Product Title
Lenz Laborglasinstrumente™ Dropping funnel, conical-shaped, with PTFE plug > Capacity (Metric): 1000mL