Learn More
VITLAB™ Dropping Pipets With Integrated Bellows
Brand:  VITLAB™ 149993
Product Code. 10081392
  • 287.00 DKK / Pack of 100
Only null left
Estimated Shipment: 04-08-2025
Specifications
5 mL
194 mm
Dropping Pipette
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
VITLAB™ Dropping Pipets With Integrated Bellows > Capacity: 5mL