Learn More
Justrite™ Drum Cabinet Vertical Two Drum Storage
Brand:  Justrite™ 8990701
Product Code. 12963425
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
2 door, self close, with drum rollers & removable, adjustable shelf, steel can accommodate pump or funnel yellow 1651mm x 1092mm x 864mm (w x d x h) 2 x 110L
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Justrite™ Drum Cabinet Vertical Two Drum Storage >