Learn More
Kimble Chase™ Drummond Microcaps Borosilicate Glass Capillary Tubes
Click to view available options
Capacity (Metric):
0.5 μL
1 μL
25 μL
50 μL
Length (Metric):
32 mm
65 mm
100 mm
Unit Size:
Pack of 100
Description
Kimble Chase Drummond Microcaps Borosilicate Glass Capillary Tubes, Capacity: 0.5 uL, Length: 32 mm, Quantity: 100/Pk.
Specifications
Specifications
Capacity (Metric) | 0.5 μL |
Material | Borosilicate Glass |
Quantity | 100/Pk. |
Type | Capillary Tube |
Length (Metric) | 32 mm |
Product Content Correction
Product Title
Kimble Chase™ Drummond Microcaps Borosilicate Glass Capillary Tubes >
Spot an opportunity for improvement?Share a Content Correction