Learn More
Drummond™ Original Portable Pipet-Aid™ Pipet Controller
Convenient alternative to mouth pipetting in the laboratory.
Brand:  Drummond™ 07897-32
4655.56 DKK valid until 2025-06-30
Use promo code "24075" to get your promotional price.
 View more versions of this product
Product Code. 11714819
  • 5445.00 DKK / Each
The Ultimate Lab Appliances Promotion
Use promo code 24075 at checkout
View Available Promotions
Description
  • Easy-to-use volume control mechanism permits precise fluid intake and delivery
  • Check valve protects against accidental fluid intake into the handle
  • Can be used with glass or plastic pipets
  • Autoclavable components
Order Info
Contact your Fisher Scientific Customer Service Representative for available replacement parts.
Certifications
CE, Euro version
Specifications
Specifications
Autoclavable
Laboratory pipetting
Pipet Controller, TC Nosepiece
Portable Filler/Dispenser
Glass or plastic pipets
220 V
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.
    Promo Code: 24075
    Expires2025-06-30
    Days
    21
    Hours
    14
    Minutes
    6
    Seconds
    53
Product Content Correction
Product Title
Drummond™ Original Portable Pipet-Aid™ Pipet Controller > 220V