missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KARTELL Drying Rack Pegs
These Drying Rack Pegs are compatible with the Kartell Draining Rack for easy air drying of laboratory equipment.
Brand: KARTELL 2130800
This item is not returnable.
View return policy
Description
- The 72 peg sockets have closed ends eliminating leakage and consequent biohazards can accomodate items with neck bore in excess of 15 mm are supplied and 11 small pegs of diameter 6 mm for small diameter items such as test tubes.
- They are easily removable for cleaning or to accomodate particularly shaped items.
- Supplied with Kartell draining rack code 213
Specifications
72 Pegs | |
Not Autoclavable | |
Flasks, Tubes | |
Cylindrical | |
Draining Rack |
Accessory | |
White | |
Polystyrene | |
Non-sterile | |
95 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
KARTELL Drying Rack Pegs > Drying Rack Pegs
Spot an opportunity for improvement?Share a Content Correction