missing translation for 'onlineSavingsMsg'
Learn More

DuP 697, Tocris Bioscience™

Product Code. 18044712
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Cyclooxygenase-2 (COX-2) inhibitor

Potent and selective inhibitor of cyclooxygenase-2 (IC50 values are 10 and 800 nM for COX-2 and COX-1 respectively). Inhibits prostaglandin synthesis and is anti-inflammatory in vivo. Orally active.

Chemical Identifiers

CAS 88149-94-4
Molecular Formula C17H12BrFO2S2
Molecular Weight (g/mol) 411.303
InChI Key AJFTZWGGHJXZOB-UHFFFAOYSA-N
Synonym mrwlzpofpa, unii-mrwlzpofpa, chembl42485, 5-bromo-2-4-fluorophenyl-3-4-methysulfonylphenyl thiophene, 5-bromo-2-4-fluorophenyl-3-4-methylsulfonylphenyl thiophene, thiophene, 5-bromo-2-4-fluorophenyl-3-4-methylsulfonyl phenyl, thiophene, 5-bromo-2-4-fluorophenyl-3-4-methylsulfonyl phenyl-cas, 5-bromo-2-4-fluorophenyl-3-4-methylsulfonyl phenyl thiophene, tocris-1430
PubChem CID 3177
ChEBI CHEBI:4720
IUPAC Name 5-bromo-2-(4-fluorophenyl)-3-(4-methylsulfonylphenyl)thiophene
SMILES CS(=O)(=O)C1=CC=C(C=C1)C2=C(SC(=C2)Br)C3=CC=C(C=C3)F

Specifications

Quantity 10 mg
Formula Weight 411.3
Percent Purity >99%
Chemical Name or Material DuP 697
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'