Få mere at vide
DuPont™ Tyvek™ IsoClean™ Sleeves
Brand: DuPont™ D14238500
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
- Sterile sleeve available in white and in one size
- Tunnelled elastication at wrist and upper arm
- This accessory is not CE certified as chemical protective clothing
- Gamma-sterilised
- Sterility assurance level (SAL) of 10-6
- Bound seams: tightly sewn seam is covered with garment fabric to reinforce seam protection and reduce potential for liquid and particle penetration
- Full traceability on all sterilized apparel with certificates of sterility available
- Tyvek™ garments are composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort
- Tyvek™ is permeable to both air and water vapour, yet repels water-based liquids and aerosols
Tekniske data
D14238500 |
Certifikater
Der kræves et partinummer for at vise resultater for certifikater. For at finde dit varenummer på tidligere ordrer, skal du bruge vores ordrestatusområde.
5 resultater fundet
Partinummer | Certifikattype | Dato | Katalognummer |
---|---|---|---|
KAS253164N | Sterilitetscertifikat | 29/09/2025 | D14238500 |
AHS2537101 | Sterilitetscertifikat | 29/09/2025 | D14238500 |
AHS2536102 | Sterilitetscertifikat | 29/09/2025 | D14238500 |
KAS253064N | Sterilitetscertifikat | 22/09/2025 | D14238500 |
KAS252864N | Sterilitetscertifikat | 22/09/2025 | D14238500 |
Korrektion af produktindhold
Produkttitel
DuPont™ Tyvek™ IsoClean™ Sleeves >
Ser du en mulighed for forbedring?Del en indholdskorrektion