Learn More
Lenz Laborglasinstrumente™ Adapters, Bent
Provides for quick and easy leak-tight connections to apparatus in the laboratory. Lenz Laborglasinstrumente™ Adapters, Bent are manufactured from durable DURAN™ borosilicate glass.
Brand: Lenz Laborglasinstrumente™ 05095004
This item is not returnable.
View return policy
Description
- With NS cone and bent glass olive at a 90° angle
- Corrosion-free and resistant to thermal shock
- With conical joint, featuring high taper accuracy and excellent tightness
Specifications
Duran | |
Adapter | |
With NS cone and bent glass olive (angle 90°) |