Learn More
Lenz Laborglasinstrumente™ Condensers Acc. to Allihn
Facilitates the condensing of vapors in the laboratory. Lenz Laborglasinstrumente™ Condensers Acc. to Allihn are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 05223211
 View more versions of this product
Product Code. 10324902
  • 576.00 DKK / Each
Estimated Shipment: 16-07-2025
Explore more special offers
Description
  • Acc. to DIN 12581
  • With NS socket and cone
  • Corrosion-free and resistant to thermal shock
  • Available with either glass olive or GL plastic olives/threads
Specifications
Specifications
For Condensing Vapors
1 Pack
Duran Glass
160 mm
14/23
Allihn Condenser
Product Title
Lenz Laborglasinstrumente™ Condensers Acc. to Allihn > Neck Size: 14/23, Jacket Length: 160mm