Learn More
Lenz Laborglasinstrumente™ Three-Neck Round Bottom Flasks, Side Necks Parallel
Especially made for distillation, reactions and to make solutions or hold, contain and collect chemicals. Lenz Laborglasinstrumente™ Three-Neck Round Bottom Flasks, Side Necks Parallel are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 03304137
 View more versions of this product
Product Code. 10599421
  • 255.00 DKK / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Acc. to DIN 12392
  • Two parallel side necks at 90° angle
  • With conical ground joints, featuring high taper accuracy and excellent tightness
  • With inscription label
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
100 mL
Borosilicate Glass, Duran Glass
14/23 NS Side Neck, 29/32 NS Center Neck
Round