Learn More
Hirschmann™ DURAN™ Class AS Micro Burettes Blue Graduations
Excellent for dispensing measured amounts of chemical solutions in the laboratory. Manufactured from durable DURAN™ glass tubing.
Brand: Hirschmann™ 3300160
Description
- Schellbach
- Bang pattern
- Main point ring graduation
- With blue graduation
- Acc. to DIN EN ISO 385
- Support base
- Conformity certified, with dated batch identification
Specifications
10 mL | |
0.02 mL | |
Glass | |
Straight | |
±0.02 mL | |
Class AS |
Borosilicate Glass | |
Bang, Schellbach Stripes | |
Blue | |
Glass | |
Microburet | |
DIN EN ISO 385 |