Learn More
DWK Life Sciences DURAN™ Coil Distillate Condenser, with 2 standard ground joints, and 2 screw-on plastic hose connections
The coil condenser serves to distill a reaction mixture or to cool a condensate.
911.00 DKK
Specifications
Material | Borosilicate Glass (3.3), PBT, Polypropylene, Silicone |
---|---|
For Use With (Application) | Condensate cooling; Product condenser for distillate separations; Distillation |
Quantity | 1 Ea. |
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | ||||||
10528821
|
DWK Life Sciences
242537105 |
NS 24/29 |
911.00 DKK
Each |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||||
10080011
|
DWK Life Sciences
242537208 |
NS 29/32 |
911.00 DKK
Each |
In Stock
Log in to see stock. |
||||||
Description
Typical for the coil condenser is its spiral riser tube, which is located in a jacket and thus washed around by the cooling medium (contrary to the Dimroth cooler). Due to this condensate flow, the condensation performance is very low although the cooling section is very long. This type of condenser is therefore usually used as a downstream additional condenser and not as an independent distillation condenser. Due to the better drainage of condensate in the convoluted riser tube, the condenser must be installed in a vertical position.
- Usually used as a downstream additional cooler due to long cooling section
- Two removable plastic olives for inlet and outlet of the cooling medium
- Tight closure due to standard ground joint
Specifications
Borosilicate Glass (3.3), PBT, Polypropylene, Silicone | |
Condensate cooling; Product condenser for distillate separations; Distillation | |
1 Ea. |