Learn More
Lenz Laborglasinstrumente™ Duran™ Glass Spiral Condensers
Brand:  Lenz Laborglasinstrumente™ 05233201
 View more versions of this product
Product Code. 10111571
  • 958.00 DKK / Each
Estimated Shipment: 17-06-2025
Explore more special offers
Specifications
For cooling condensates
1 Each
Duran Glass
160 mm
14/23
Spiral Condenser
Product Title
Lenz Laborglasinstrumente™ Duran™ Glass Spiral Condensers > Jacket Length: 160mm