Learn More
Lenz Laborglasinstrumente™ Duran™ Sprayer Heads
332.00 DKK - 426.00 DKK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
10509751
|
Lenz Laborglasinstrumente™
05470004 |
100 mL |
332.00 DKK
Each |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
11973531
|
Lenz Laborglasinstrumente™
05470605 |
6 mL |
415.00 DKK
Each |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
11516914
|
Lenz Laborglasinstrumente™
05471205 |
12 mL |
426.00 DKK
Each |
Estimated Shipment: 29-08-2025
Log in to see stock. |
||||||
Specifications
Duran Glass | |
1/Pk. | |
Sprayer Head |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Duran™ Sprayer Heads