Learn More
Macherey-Nagel™ Durasil-25 TLC Plate
Shop All Macherey-Nagel Products
Click to view available options
Dimensions (L x W):
10 x 20 cm
20 x 20 cm
Unit Size:
Pack of 25
Pack of 50
Specifications
Specifications
Material | Durasil-25 |
Particle Size | 5 to 17 μm |
Pore Size | 60 Å |
Quantity | 25 Pk. |
Type | TLC Plate |
Pore Volume | 0.75 mL/g |
Specific Surface | ∼500 m2/g |
Thickness (Metric) | 0.25 mm |
Dimensions (L x W) | 20 x 20 cm |
Product Content Correction
Product Title
Macherey-Nagel™ Durasil-25 TLC Plate >
Spot an opportunity for improvement?Share a Content Correction