missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ DYRK2 Recombinant Protein
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | AAH05809 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Kinase Assay, Western Blot |
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
Gene ID (Entrez) | 8445 |
Molecular Weight (g/mol) | 83.82 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16122462
|
Abnova™
H00008445-P01.10ug |
10 ug |
2665.00 DKK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16132462
|
Abnova™
H00008445-P01.25ug |
25 ug |
4040.00 DKK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH05809 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
83.82 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DYRK2 | |
Human | |
Recombinant | |
Solution |
Antibody Production, Protein Array, ELISA, Kinase Assay, Western Blot | |
8445 | |
DYRK2 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MNDHLHVGSHAHGQIQVQQLFEDNSNKRTVLTTQPNGLTTVGKTGLPVVPERQLDSIHRRQGSSTSLKSMEGMGKVKATPMTPEQAMKQYMQKLTAFEHHEIFSYPEIYFLGLNAKKRQGMTGGPNNGGYDDDQGSYVQVPHDHVAYRYEVLKVIGKGSFGQVVKAYDHKVHQHVALKMVRNEKRFHRQAAEEIRILEHLRKQDKDNTMNVIHMLENFTFRNHICMTFELLSMNLYELIKKNKFQGFSLPLVRKFAHSILQCLDALHKNRIIHCDLKPENILLKQQGRSGIKVIDFGSSCYEHQRVYTYIQSRFYRAPEVILGARYGMPIDMWSLGCILAELLTGYPLLPGEDEGDQLACMIELLGMPSQKLLDASKRAKNFVSSKGYPRYCTVTTLSDGSVVLNGGRSRRGKLRGPPESREWGNALKGCDDPLFLDFLKQCLEWDPAVRMTPGQALRHPWLRRRLPKPPTGEKTSVKRITESTGAITSISKLPPPSSSASKLRTNLAQMTDANGNIQQRTVLPKLVS | |
FLJ21217/FLJ21365 | |
DYRK2 | |
Wheat Germ (in vitro) | |
GST |