Description
- Easy to use earplugs that provide effective protection, for low to moderate noise exposure
- They require no roll down, simply seals the ear canal entrance to achieve a secure fit
- Made from soft, comfortable, slow recovery foam
Specifications
Specifications
Color | Yellow |
Certifications/Compliance | CE Approved |
Pairs per Package | 1000 Pairs/Box |
Noise Reduction Rating (NRR) | 21 dB |
Corded | Uncorded |
Product Type | Ear Plugs |
Dielectric | No |
Material | Slow-recovery Polyurethane Foam |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Product Title
3M™ E-A-R™ E-A-Rsoft™ 21 Earplugs >
Spot an opportunity for improvement?Share a Content Correction