missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E-Tag Epitope Tag Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
1860.00 DKK - 5125.00 DKK
Specifications
Antigen | E-Tag Epitope Tag |
---|---|
Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:300-1:10000, Immunoprecipitation 0.5-2 μg/mg of lysate |
Applications | Western Blot, Immunofluorescence, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18640506
|
Novus Biologicals
NBP3-05892-100ul |
100 μg |
2995.00 DKK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18691726
|
Novus Biologicals
NBP3-05892-500ul |
500 μg |
5125.00 DKK
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18692756
|
Novus Biologicals
NBP3-05892-50ul |
50 μg |
1860.00 DKK
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
E-Tag Epitope Tag Polyclonal antibody specifically detects E-Tag Epitope Tag in Epitope Tag samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
E-Tag Epitope Tag | |
Western Blot, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Epitope Tags | |
0.2 um filtered solution in PBS | |
Produced in rabbits immunized with A synthetic peptide corresponding to the E-Tag Epitope Tag sequence (GAPVPYPDPLEPR). | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:300-1:10000, Immunoprecipitation 0.5-2 μg/mg of lysate | |
Polyclonal | |
Purified | |
RUO | |
Tag | |
E epitope tag, GAPVPYPDPLEPR epitope tag, GAPVPYPDPLEPR tag | |
IgG | |
Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
E-Tag Epitope Tag Antibody, Novus Biologicals™