Få mere at vide
Contec™ EasySat Mop
Designed for the cleanroom with a built-in 52 ounce solution tank that completely eliminates the need for a bucket and wringer. Contec™ EasySat Mop is an effective and quick solution to your cleaning challenges.
Brand: Contec™ BLMOPE
Beskrivelse
- Hydrophilic polyester/clean ester, laser cut sealed edge foam material.
- Translucent tank allows user to see solution
- Fully autoclavable for critical arease
- Easy to use
- Compatible with a wide range of solvents
Tekniske data
55 x 46 cm | |
Bucketless Mop | |
Non-sterile | |
Autoclavable |
Certifikater
Der kræves et partinummer for at vise resultater for certifikater. For at finde dit varenummer på tidligere ordrer, skal du bruge vores ordrestatusområde.
Partinummer | Certifikattype | Dato | Katalognummer |
---|---|---|---|
126810-220727 | Kvalitetscertifikat | 12/01/2023 | BLMOPE |
126422-220705 | Kvalitetscertifikat | 08/10/2022 | BLMOPE |
210321 | Overensstemmelsescertifikat | 29/07/2021 | BLMOPE |