Learn More
Thermo Scientific™ EasyStrip™ Plus Tube Strip with Attached Ultra Clear Caps, Sustain™ Series
The Thermo Scientific EasyStrip Plus Tube Strip with Attached Ultra Clear Caps, Sustain Series, is designed to reduce the risk of cross-contamination in PCR and qPCR assays.
Brand: Thermo Scientific™ AB2005SS
1139.85 DKK valid until 2025-06-30
Use promo code "24108" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
Description
The Thermo Scientific EasyStrip Plus Tube Strip with Attached Ultra Clear Caps, Sustain Series, is designed to reduce the risk of cross-contamination in PCR and qPCR assays. The tube strip is compatible with 0.2 mL thermal cycler blocks and features individually attached flat caps for quick and easy access to individual samples. In addition, graduated 20 ÎĽL markers on each tube enable visual volume verification and measurement of reagent and sample additions.
Features and benefits
- Suitable for qPCR and PCR applications
- Avoid cross contamination with the individually attached caps
- Verify tube reaction volumes with graduated 20 ÎĽL markings on each tube
- Track samples using the end tabs to label each strip
- Identify individual wells quickly and prevent pipetting errors using the labels (A-H) on each tube cap and cap hinge (visible when cap is open and closed)
- Certified DNase-, RNase-, human DNA-, and PCR inhibitor-free
A breakthrough in sustainable PCR technology
Thermo Scientific Sustain Series PCR plastics signify a breakthrough in our PCR and quantitative PCR (qPCR) product offering that goes beyond providing accurate and reproducible PCR data. They represent our commitment to integrating more sustainable practices into our manufacturing processes, without compromising on product quality or performance. Our Sustain Series PCR plastics are manufactured under ISO 9001 guidelines within a Class 100,000 cleanroom from a biobased medical-grade virgin polypropylene that is chemically and molecularly identical to our existing Thermo Scientific PCR plastics. These products are supported by certification from the globally recognized International Sustainability and Carbon Certification (ISCC) system. This greener approach is equivalent to 1.96X lower CO2 equivalent emissions per kg of biobased polypropylene resin compared to traditional fossil fuel-based polypropylene resin.
Additional product details
- Uniform, ultra-thin walls enable maximum and consistent heat transfer for equally high performance of every sample
- Each well is visually inspected and electrostatically tested for defects and contaminants, and every lot is run through a PCR cycling test to evaluate sealing
- Manufactured from second generation bio-circular feedstocks that are chemically and molecularly identical to our existing Thermo Scientific PCR plastics
Specifications
Certified DNase-, RNase-, Human DNA-, and PCR Inhibitor-free | |
250 Strips | |
Optical (Attached) | |
Yes |
Natural | |
Polypropylene | |
Autoclavable | |
Less waste, renewable material |