Learn More
Invitrogen™ eBioscience™ Phytohemagglutinin-L (PHA-L) Solution (500X)
The Phytohemagglutinin-L (PHA-L) Solution (500X) is a ready to use solution of PHA-L in aqueous buffer.
Brand: Invitrogen™ 00-4977-03
This item is not returnable.
View return policy
Description
- A ready to use solution of PHA-L in aqueous buffer
- PHA is a mixture of lectins isolated from the red kidney bean, Phaseolus vulgaris
- It is the fraction of PHA that exhibits strong leukoagglutinating and mitogenic activity, but very little erythroagglutinating activity
- This reagent is intended for use in in vitro activation of human and mouse leukocytes
Specifications
4 x 100 μL | |
eBioscience™ | |
Phytohemagglutinin-L (PHA-L) Solution |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Technical Data Sheet: Phytohemagglutinin-L (PHA-L) Solution (500X)
Version: 14 March 2017 Rev. 10
File size: 251.6 kB
Product Content Correction
Product Title
Invitrogen™ eBioscience™ Phytohemagglutinin-L (PHA-L) Solution (500X) > 4 x 100 μL