Learn More
Invitrogen™ EDAC, 1-Ethyl-3-(3-Dimethylaminopropyl)carbodiimide, Hydrochloride
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Protocols for crosslinking molecules with this reagent can be found in Bioconjugate Techniques, by G.T. Hermanson (B-7884).
Specifications
Specifications
Quantity | 100 mg |
Molecular Weight (g/mol) | 191.7 |
Physical Form | Solid |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Product Title
Invitrogen™ EDAC, 1-Ethyl-3-(3-Dimethylaminopropyl)carbodiimide, Hydrochloride >
Spot an opportunity for improvement?Share a Content Correction