missing translation for 'onlineSavingsMsg'
Learn More

EIF4EBP1, Rabbit anti-Human, PE, Clone: A5, Abnova™

Rabbit Monoclonal Antibody

Brand:  Abnova MAB18771.100Reactions

Product Code. 16120150

  • 4965.00 DKK / 100 reactions

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

EIF4EBP1
Monoclonal
PE
In PBS, pH 7.4 (0.2% BSA, 0.09% sodium azide).
4E-BP1/4EBP1/BP-1/MGC4316/PHAS-I
Rabbit
100 Reactions
Primary
Human, Mouse, Rat
Antibody
IgG1 Îş
Flow Cytometry
A5
Flow Cytometry (5 uL/106 cells) The optimal working dilution should be determined by the end user.
eukaryotic translation initiation factor 4E binding protein 1
EIF4EBP1
A synthetic phosphopeptide corresponding to residues surrounding T37/T46 of human EIF4EBP1.
RUO
1978
Store at 4°C.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
EIF4EBP1, Rabbit anti-Human, PE, Clone: A5, Abnova™ > 100 Reactions; PE

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.