Learn More
Invitrogen™ EK-Away™ Resin
Specifically designed for removal of EKMaxor other enterokinase enzymes following cleavage of proteins containing the enterokinase cleavage site
5860.00 DKK
Specifications
Content And Storage | EK-Away™ is provided as a 50% ethanol slurry and should be stored at +4°C. 10X binding and 10X stripping buffers are included. 7.5 ml resin will remove 250 units of enterokinase; 30 ml will remove 1000 units of EKMax™. All reagents are guaranteed stable for 6 months when stored properly. |
---|---|
Form | Liquid Suspension |
Product Line | EK-Away™ |
Type | Resin |
Stationary Phase | Enterokinase |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10403982
|
Invitrogen™
R18001 |
7.5 mL |
5860.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The resin is conjugated with soybean trypsin inhibitor, which has a high affinity and binding capacity for enterokinase
- The enzyme catalytic site binds this agarose-based resin for simple batch or column-bound removal of EKMaxor other enterokinase preparations
GST-Tagged Protein Purification, His-Tagged Protein Purification, Protein and Peptide Purification, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis
Specifications
EK-Away™ is provided as a 50% ethanol slurry and should be stored at +4°C. 10X binding and 10X stripping buffers are included. 7.5 ml resin will remove 250 units of enterokinase; 30 ml will remove 1000 units of EKMax™. All reagents are guaranteed stable for 6 months when stored properly. | |
EK-Away™ | |
Enterokinase |
Liquid Suspension | |
Resin | |
Affinity |