Learn More
Mettler Toledo™ Electrode Arm, EasyPlace Complete
The EasyPlace electrode arm supports efficient measurement and calibration. With perfect vertical movement, spillage risks are minimized. Includes parking beaker, parking beaker insert and sachet holder.
Brand: Mettler Toledo™ 30671569
1992.08 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Precise Control
- Perfectly vertical sensor movement makes it easy to insert the sensor into samples
- The right place
- The EasyPlace contains dedicated positions for calibration, short- and long term sensor storage and measurement, simplifying everyday workflows
- Fits right in
- The EasyPlace electrode arm fits together with the EasyMix magnetic stirrer and standard sachets, simplifying operation and saving space
Specifications
Electrode Arm, EasyPlace Complete | |
EasyPlace |
Promotions
-
Multiple Manufacturers
Focus On Clean Water
Shop now for up to 60% off Clean Water Research essentialsPromo Code: 24139Expires: 2025-08-21
New Offer Save Now