Learn More
Thermo Scientific™ Electroporation Cuvettes
GREENER_CHOICE
Increase the ease of sample identification and labelling with these premium-grade polycarbonate electroporation cuvettes, which have frosted, colored caps.
Brand:  Thermo Scientific™ 5510-11
1590.69 DKK valid until 2025-08-24
Use promo code "24117" to get your promotional price.
 View more versions of this product
Product Code. 12338212
  • 1780.00 DKK / Pack of 50
Spotlight
Use promo code 24157 at checkout
View Available Promotions
Only null left
Estimated Shipment: 05-08-2025
Description
Increase the ease of sample identification and labeling with premium-grade polycarbonate Thermo Scientific™ Electroporation Cuvettes, which have frosted, colored caps.
  • Available in 1mm, 2mm, or 4mm gap sizes
  • Pre-sterilized and individually wrapped
  • Compatible with all common electroporators
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Retort Ring
1mm gap
Stainless Steel
50 mm
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24157
    Expires2025-08-31
    Days
    45
    Hours
    6
    Minutes
    37
    Seconds
    5
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/07/2025 and 31/08/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
Product Content Correction
Product Title
Thermo Scientific™ Electroporation Cuvettes > 1mm