Learn More
Thermo Scientific™ Electroporation Cuvettes
GREENER_CHOICE
Increase the ease of sample identification and labelling with these premium-grade polycarbonate electroporation cuvettes, which have frosted, colored caps.
Brand:  Thermo Scientific™ 5540-11
1226.89 DKK valid until 2025-08-24
Use promo code "24117" to get your promotional price.
 View more versions of this product
Product Code. 12348212
  • 1985.00 DKK / Pack of 50
Only null left
Estimated Shipment: 05-08-2025
Description
Increase the ease of sample identification and labeling with premium-grade polycarbonate Thermo Scientific™ Electroporation Cuvettes, which have frosted, colored caps.
  • Available in 1mm, 2mm, or 4mm gap sizes
  • Pre-sterilized and individually wrapped
  • Compatible with all common electroporators
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sterile
Electroporation Cuvet
4mm gap
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Electroporation Cuvettes > 4mm